2-(1H-1,2,4-Triazol-1-yl)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
OR14080-10G
OR14080-5G
APOSOR14080-10GEA
275
EUR
APOSOR14080-10G
APOSOR14080-5G
2-(1H-1,2,4-Triazol-1-yl)benzaldehyd
2-(1H-1,2,4-Triazol-1-yl)benzaldehyd
Formel:
C₉H₇N₃O Molecular Weight: 173,1714 g/mol Schmelzpunkt: 110…113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03617913 CAS-Nummer: 138479-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...