[5-Amino-2-(4-chlorophenoxy)phenyl]methanol

Lieferant: Apollo Scientific

Synonyme: 5-Amino-2-(4-chlorophenoxy)benzyl alcohol

OR14087-10G OR14087-1G OR14087-5G
APOSOR14087-10GEA 1690 EUR
APOSOR14087-10G APOSOR14087-1G APOSOR14087-5G
[5-Amino-2-(4-chlorophenoxy)phenyl]methanol
[5-Amino-2-(4-chlorophenoxy)phenyl]methanol
Formel: C₁₃H₁₂ClNO₂
Molecular Weight: 249,7 g/mol
Schmelzpunkt: 129…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689719
CAS-Nummer: 924869-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR