5-Bromo-6-phenyl-3(2H)-pyridazinone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-6-phenylpyridazin-3(2H)-one
OR14091-10G
OR14091-1G
OR14091-5G
APOSOR14091-10GEA
134
EUR
APOSOR14091-10G
APOSOR14091-1G
APOSOR14091-5G
5-Bromo-6-phenyl-3(2H)-pyridazinone
5-Bromo-6-phenyl-3(2H)-pyridazinone
Formel:
C₁₀H₇BrN₂O Molecular Weight: 251,09 g/mol Schmelzpunkt: 234…237 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02083298 CAS-Nummer: 90766-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...