3-Chlor-5-nitro-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR14094-10G
OR14094-1G
OR14094-5G
APOSOR14094-10GEA
62.5
EUR
APOSOR14094-10G
APOSOR14094-1G
APOSOR14094-5G
3-Chlor-5-nitro-1H-indazol
3-Chloro-5-nitro-1H-indazole
Formel:
C₇H₄ClN₃O₂ Molecular Weight: 197,5800 g/mol Schmelzpunkt: 218…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005692 CAS-Nummer: 4812-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...