2-(2-Chloro-3-pyridinyl)-1,3-benzoxazole
Lieferant: Apollo Scientific
OR14107-1G
APOSOR14107-1GEA
538
EUR
APOSOR14107-1G
2-(2-Chloro-3-pyridinyl)-1,3-benzoxazole
2-(2-Chloro-3-pyridinyl)-1,3-benzoxazole
Formel:
C₁₂H₇ClN₂O Molecular Weight: 230,66 g/mol Schmelzpunkt: 94…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689672 CAS-Nummer: 134673-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...