6,8-Dichloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Lieferant: Apollo Scientific
Synonyme:
6,8-Dichloro-2,3-dihydro-3-phenyl-2-thioxo-1H-quinazolin-4-one
OR14110-1G
OR14110-5G
APOSOR14110-1GEA
180
EUR
APOSOR14110-1G
APOSOR14110-5G
6,8-Dichloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
6,8-Dichloro-3-phenyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone
Formel:
C₁₄H₈Cl₂N₂OS Molecular Weight: 323,2 g/mol Schmelzpunkt: 274…275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689694 CAS-Nummer: 67867-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...