tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate

Lieferant: Apollo Scientific

Synonyme: 4-{2-[4-(Hydrazinocarbonyl)piperidin-1-yl]ethyl}piperazine, N1-BOC protected

OR14125-1G OR14125-5G
APOSOR14125-1GEA 538 EUR
APOSOR14125-1G APOSOR14125-5G
tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
tert-Butyl-4-{2-[4-(hydrazinocarbonyl)piperidino]ethyl}tetrahydro-1(2H)-pyrazinecarboxylate
Formel: C₁₇H₃₃N₅O₃
Molecular Weight: 355,48 g/mol
MDL: MFCD06808601
CAS-Nummer: 874831-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR