Methyl-4-amino-3-hydroxybenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-amino-3-hydroxybenzoate

OR14134-25G OR14134-5G OR14134-1G
APOSOR14134-25GEA 87 EUR
APOSOR14134-25G APOSOR14134-5G APOSOR14134-1G
Methyl-4-amino-3-hydroxybenzoat
Methyl-4-amino-3-hydroxybenzoat
Formel: C₈H₉NO₃
Molecular Weight: 167,17 g/mol
Schmelzpunkt: 118…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017093
CAS-Nummer: 63435-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR