Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

OR14137-1G OR14137-500MG OR14137-5G
APOSOR14137-1GEA 174 EUR
APOSOR14137-1G APOSOR14137-500MG APOSOR14137-5G
Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate
Methyl-2-(chloromethyl)-1,3-benzoxazole-5-carboxylate
Formel: C₁₀H₈ClNO₃
Molecular Weight: 225.63 g/mol
Schmelzpunkt: 90…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689722
CAS-Nummer: 924862-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR