Methyl-3-aminoisonicotinat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-aminoisonicotinate, Methyl 3-aminopyridine-4-carboxylate
OR14139-25G
OR14139-5G
OR14139-100G
APOSOR14139-25GEA
114
EUR
APOSOR14139-25G
APOSOR14139-5G
APOSOR14139-100G
Methyl-3-aminoisonicotinat
Methyl-3-aminoisonicotinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.20 g/mol Schmelzpunkt: 84…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082535 CAS-Nummer: 55279-30-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...