Methyl-4-(3-amino-2-pyridinyl)benzenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-(3-aminopyridin-2-yl)benzoate
OR14140-100MG
OR14140-250MG
APOSOR14140-100MGEA
31.9
EUR
APOSOR14140-100MG
APOSOR14140-250MG
Methyl-4-(3-amino-2-pyridinyl)benzenecarboxylate
Methyl-4-(3-amino-2-pyridinyl)benzenecarboxylate
Formel:
C₁₃H₁₂N₂O₂ Schmelzpunkt: 135…137 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689702 CAS-Nummer: 924869-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...