Methyl-1,3-benzoxazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl benzo[d]oxazole-5-carboxylate
OR14141-1G
OR14141-5G
OR14141-10G
APOSOR14141-1GEA
102
EUR
APOSOR14141-1G
APOSOR14141-5G
APOSOR14141-10G
Methyl-1,3-benzoxazole-5-carboxylate
Methyl-1,3-benzoxazole-5-carboxylate
Formel:
C₉H₇NO₃ Schmelzpunkt: 110…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689687 CAS-Nummer: 924869-17-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...