4-Methyl-7-nitro-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR14147-1G OR14147-250MG OR14147-5G
APOSOR14147-1GEA 60.9 EUR
APOSOR14147-1G APOSOR14147-250MG APOSOR14147-5G
4-Methyl-7-nitro-1H-indazol
4-Methyl-7-nitro-1H-indazole
Formel: C₈H₇N₃O₂
Molecular Weight: 177,1601 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689688
CAS-Nummer: 104103-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR