1-[(6-Chloro-3-pyridinyl)methyl]-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR14203-1G OR14203-250MG
APOSOR14203-1GEA 49.3 EUR
APOSOR14203-1G APOSOR14203-250MG
1-[(6-Chloro-3-pyridinyl)methyl]-1H-indole-3-carbaldehyde
1-[(6-Chloro-3-pyridinyl)methyl]-1H-indole-3-carbaldehyde
Formel: C₁₅H₁₁ClN₂O
Molecular Weight: 270,72 g/mol
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200974
CAS-Nummer: 886361-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR