2-(4-Formylphenyl)isonicotinonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR14211-1G
APOSOR14211-1GEA
400
EUR
APOSOR14211-1G
2-(4-Formylphenyl)isonicotinonitril 95%
2-(4-Formylphenyl)isonicotinonitril
Formel:
C₁₃H₈N₂O Schmelzpunkt: 175…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200959 CAS-Nummer: 253801-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...