2-Iod-p-toluidin
Lieferant: Apollo Scientific
|
Gefahr
|
OR14214-5G
OR14214-1G
OR14214-25G
APOSOR14214-5GEA
29
EUR
APOSOR14214-5G
APOSOR14214-1G
APOSOR14214-25G
2-Iod-p-toluidin
2-Iod-p-toluidin
Formel:
C₇H₈IN Molecular Weight: 233,05 g/mol Schmelzpunkt: 39…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407391 CAS-Nummer: 29289-13-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...