6-Phenyl-3-pyridinamine 95+%

Lieferant: Apollo Scientific
Achtung

OR14233-250MG
APOSOR14233-250MGEA 50.9 EUR
APOSOR14233-250MG
6-Phenyl-3-pyridinamine 95+%
6-Phenyl-3-pyridinamine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170,22 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114197
CAS-Nummer: 126370-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR