2-Propyl-1,3-benzoxazol-6-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Amino-2-propyl-1,3-benzoxazole
OR14234-250MG
OR14234-500MG
APOSOR14234-250MGEA
116
EUR
APOSOR14234-250MG
APOSOR14234-500MG
2-Propyl-1,3-benzoxazol-6-amine
2-Propyl-1,3-benzoxazol-6-amine
Formel:
C₁₀H₁₂N₂O Molecular Weight: 176,22 g/mol Schmelzpunkt: 83…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200963 CAS-Nummer: 875851-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...