3-(Piperidin-1-ylsulfonyl)anilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR14248-10G
OR14248-1G
OR14248-5G
APOSOR14248-10GEA
716
EUR
APOSOR14248-10G
APOSOR14248-1G
APOSOR14248-5G
3-(Piperidin-1-ylsulfonyl)anilin 97%
3-(Piperidin-1-ylsulfonyl)anilin
Formel:
C₁₁H₁₆N₂O₂S Schmelzpunkt: 116…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02063328 CAS-Nummer: 22184-99-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...