3-(Morpholinosulfonyl)anilin 98%

Lieferant: Apollo Scientific
Achtung

OR14249-1G OR14249-5G OR14249-10G
APOSOR14249-1GEA 105 EUR
APOSOR14249-1G APOSOR14249-5G APOSOR14249-10G
3-(Morpholinosulfonyl)anilin 98%
3-(Morpholinosulfonyl)anilin
Formel: C₁₀H₁₄N₂O₃S
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02090909
CAS-Nummer: 22184-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR