5-Amino-3-(4-bromphenyl)-1H-pyrazol
Lieferant: Apollo Scientific
Synonyme:
3-(4-Bromophenyl)-1H-pyrazol-5-amine
OR14269-25G
OR14269-5G
APOSOR14269-25GEA
253
EUR
APOSOR14269-25G
APOSOR14269-5G
5-Amino-3-(4-bromphenyl)-1H-pyrazol
5-Amino-3-(4-bromphenyl)-1H-pyrazol
Formel:
C₉H₈BrN₃ Molecular Weight: 238,09 g/mol Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00558911 CAS-Nummer: 78583-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...