Amantadin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Aminotricyclo[3.3.1.1(3,7)]decan, 1-Adamantylamin, 1-Aminoadamantan
OR14310-100G
OR14310-25G
OR14310-500G
APOSOR14310-100GEA
56.7
EUR
APOSOR14310-100G
APOSOR14310-25G
APOSOR14310-500G
Amantadin 95%
Amantadin
Formel:
C₁₀H₁₇N Molecular Weight: 151,25 g/mol Schmelzpunkt: 206…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074732 CAS-Nummer: 768-94-5 EINECS: 212-201-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...