4-Aminoantipyrin 98+%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-on, 4-Aminophenazon

OR14314-100G
APOSOR14314-100GEA 60.9 EUR
APOSOR14314-100G
4-Aminoantipyrin 98+%
4-Aminoantipyrin
Formel: C₁₁H₁₃N₃O
Molecular Weight: 203,24 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003145
CAS-Nummer: 83-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR