2,2-Dimethylbernsteinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,2-Dimethyl-1,4-butanedioic acid, α,α-Dimethylsuccinic acid

OR14327-10G OR14327-25G
APOSOR14327-10GEA 21.9 EUR
APOSOR14327-10G APOSOR14327-25G
2,2-Dimethylbernsteinsäure
2,2-Dimethylbernsteinsäure
Formel: C₆H₁₀O₄
Molecular Weight: 146,14 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004196
CAS-Nummer: 597-43-3
EINECS: 209-899-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR