5-Chinolinol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Hydroxyquinoline, Quinolin-5-ol
OR14331-5G
OR14331-10G
OR14331-25G
APOSOR14331-5GEA
58
EUR
APOSOR14331-5G
APOSOR14331-10G
APOSOR14331-25G
5-Chinolinol 98%
5-Chinolinol
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 223…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006792 CAS-Nummer: 578-67-6 EINECS: 209-428-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...