6-Chinolinol
Lieferant: Apollo Scientific
|
Achtung
|
OR14332-100G
OR14332-25G
OR14332-5G
APOSOR14332-100GEA
0
EUR
APOSOR14332-100G
APOSOR14332-25G
APOSOR14332-5G
6-Chinolinol
6-Chinolinol
Formel:
C₉H₇NO Schmelzpunkt: 188…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047611 CAS-Nummer: 580-16-5 EINECS: 209-454-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...