3-Noradamantanecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR14339-5GEA
125
EUR
APOSOR14339-5G
APOSOR14339-250MG
APOSOR14339-1G
3-Noradamantanecarboxylic acid
3-Noradamantanecarboxylic acid
Formel:
C₁₀H₁₄O₂ Molecular Weight: 166,22 g/mol Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00213463 CAS-Nummer: 16200-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...