4-Iod-5-methylisoxazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR14344-1G
OR14344-250MG
APOSOR14344-1GEA
241
EUR
APOSOR14344-1G
APOSOR14344-250MG
4-Iod-5-methylisoxazol 97%
4-Iodo-5-methylisoxazole
Formel:
C₄H₄INO Molecular Weight: 208,985 g/mol Schmelzpunkt: 27…28 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06411541 CAS-Nummer: 7064-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...