2,5-Dimethyl-1,3-oxazole-4-carboxylic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dimethyloxazole-4-carboxylic acid
OR14345-100MG
OR14345-1G
OR14345-250MG
APOSOR14345-100MGEA
21.9
EUR
APOSOR14345-100MG
APOSOR14345-1G
APOSOR14345-250MG
2,5-Dimethyl-1,3-oxazole-4-carboxylic acid 98%
2,5-Dimethyl-1,3-oxazole-4-carboxylic acid
Formel:
C₆H₇NO₃ Molecular Weight: 141.13 g/mol Schmelzpunkt: 232…233 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03011595 CAS-Nummer: 23000-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...