(4-Aminophenyl)[4-(2-pyridinyl)piperazino]methanone
Lieferant: Apollo Scientific
|
Achtung
|
OR14362-1G
OR14362-250MG
APOSOR14362-1GEA
74.1
EUR
APOSOR14362-1G
APOSOR14362-250MG
(4-Aminophenyl)[4-(2-pyridinyl)piperazino]methanone
(4-Aminophenyl)[4-(2-pyridinyl)piperazino]methanone
Formel:
C₁₆H₁₈N₄O Molecular Weight: 282,35 g/mol Schmelzpunkt: 138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659555 CAS-Nummer: 885949-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...