3'-Chlor-4'-hydroxyacetophenon

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(3-Chlor-4-hydroxyphenyl)ethan-1-on, 1-(3-Chlor-4-hydroxyphenyl)ethanon

OR14369-1G OR14369-5G OR14369-25G
APOSOR14369-1GEA 21.9 EUR
APOSOR14369-1G APOSOR14369-5G APOSOR14369-25G
3'-Chlor-4'-hydroxyacetophenon
3'-Chlor-4'-hydroxyacetophenon
Formel: C₈H₇ClO₂
Molecular Weight: 170,6 g/mol
Schmelzpunkt: 92…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00663621
CAS-Nummer: 2892-29-7
UN: 3335
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR