3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(1,3,4,5,6,7-Hexahydro-3-oxo-2H-indazol-2-yl)benzoic acid

OR14390-1G OR14390-500MG
APOSOR14390-1GEA 36.4 EUR
APOSOR14390-1G APOSOR14390-500MG
3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid
3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid
Formel: C₁₄H₁₄N₂O₃
Molecular Weight: 258,28 g/mol
Schmelzpunkt: 281 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659578
CAS-Nummer: 885949-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR