6-(1H-Pyrrol-1-yl)nicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR14393-1G
OR14393-250MG
APOSOR14393-1GEA
0
EUR
APOSOR14393-1G
APOSOR14393-250MG
6-(1H-Pyrrol-1-yl)nicotinic acid
6-(1H-Pyrrol-1-yl)nicotinic acid
Formel:
C₁₀H₈N₂O₂ Schmelzpunkt: 289…291 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067843 CAS-Nummer: 175135-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...