7-Methoxyindol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Methoxy-1H-indole, Indol-7-yl methyl ether
OR1441-5G
OR1441-25G
APOSOR1441-5GEA
56.7
EUR
APOSOR1441-5G
APOSOR1441-25G
7-Methoxyindol 98%
7-Methoxyindol
Formel:
C₉H₉NO Molecular Weight: 147.18 g/mol Siedepunkt: 108…110 °C/0.3mmHg Dichte: 1.126 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00047203 CAS-Nummer: 3189-22-8 EINECS: 221-690-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...