Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4H-thieno[3,2-b]pyrrole-5-carboxylate

OR1442-1G OR1442-5G
APOSOR1442-1GEA 116 EUR
APOSOR1442-1G APOSOR1442-5G
Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate 98%
Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylate
Formel: C₈H₇NO₂S
Molecular Weight: 181,2117 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06797484
CAS-Nummer: 82782-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR