3-Hydroxy-4-nitrobenzoesäuremethylester 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl-3-hydroxy-4-nitrobenzoat
OR14421-5G
OR14421-25G
APOSOR14421-5GEA
21.9
EUR
APOSOR14421-5G
APOSOR14421-25G
3-Hydroxy-4-nitrobenzoesäuremethylester 97%
Methyl-3-hydroxy-4-nitrobenzoate
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol Schmelzpunkt: 92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016997 CAS-Nummer: 713-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...