2-Amino-5-ethylthio-1,3,4-thiadiazole

Lieferant: Apollo Scientific
Achtung

OR14426-25G OR14426-5G
APOSOR14426-25GEA 299 EUR
APOSOR14426-25G APOSOR14426-5G
2-Amino-5-ethylthio-1,3,4-thiadiazole
2-Amino-5-ethylthio-1,3,4-thiadiazole
Formel: C₄H₇N₃S₂
Molecular Weight: 161,25 g/mol
Schmelzpunkt: 133…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040401
CAS-Nummer: 25660-70-2
EINECS: 247-169-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR