4-Isobutyl-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulphide 95%

Lieferant: Apollo Scientific

Synonyme: 4-Isobutyl-5-phenyl-4H-1,2,4-triazole-3-thiol

OR14462-1G
APOSOR14462-1GEA 180 EUR
APOSOR14462-1G
4-Isobutyl-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulphide 95%
4-Isobutyl-5-phenyl-4H-1,2,4-triazol-3-ylhydrosulphide
Formel: C₁₂H₁₅N₃S
Molecular Weight: 233,34 g/mol
Schmelzpunkt: 187…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05134110
CAS-Nummer: 26029-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR