Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 3,5-dichloro-4-methoxybenzoate
OR14469-10G
OR14469-1G
OR14469-5G
APOSOR14469-10GEA
894
EUR
APOSOR14469-10G
APOSOR14469-1G
APOSOR14469-5G
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Methyl-3,5-dichloro-4-methoxybenzenecarboxylate
Formel:
C₉H₈Cl₂O₃ Molecular Weight: 235,07 g/mol Schmelzpunkt: 74…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017189 CAS-Nummer: 24295-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...