4-Butyl-5-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one 95%

Lieferant: Apollo Scientific
Achtung

OR14475-250MG
APOSOR14475-250MGEA 36.4 EUR
APOSOR14475-250MG
4-Butyl-5-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one 95%
4-Butyl-5-phenyl-2,4-dihydro-3H-1,2,4-triazol-3-one
Formel: C₁₂H₁₅N₃O
Molecular Weight: 217,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00098834
CAS-Nummer: 271798-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR