Furan-2-sulphonyl chloride 97+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Furan-2-sulphonylchloride
OR14486-1G
OR14486-250MG
APOSOR14486-1GEA
493
EUR
APOSOR14486-1G
APOSOR14486-250MG
Furan-2-sulphonyl chloride 97+%
Furan-2-sulphonyl chloride
Formel:
C₄H₃ClO₃S Molecular Weight: 166,58 g/mol Siedepunkt: 79…81 °C/10mmHg Lagertemperatur: Gefrierschrank |
MDL:
MFCD06658968 CAS-Nummer: 52665-48-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...