Ethyl-5-(2-furyl)isoxazole-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 5-(furan-2-yl)isoxazole-3-carboxylate

OR1450-100MG OR1450-250MG OR1450-500MG OR1450-2.5G OR1450-5G
APOSOR1450-100MGEA 61.1 EUR
APOSOR1450-100MG APOSOR1450-250MG APOSOR1450-500MG APOSOR1450-2.5G APOSOR1450-5G
Ethyl-5-(2-furyl)isoxazole-3-carboxylate
Ethyl-5-(2-furyl)isoxazole-3-carboxylate
Formel: C₁₀H₉NO₄
Molecular Weight: 207,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797477
CAS-Nummer: 33545-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR