3,4-Dihydro-2H-chromen-2-ylmethanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3,4-Dihydro-2H-chromen-2-yl)methanol, 2-(Hydroxymethyl)chroman
OR14501-1G
OR14501-250MG
APOSOR14501-1GEA
249
EUR
APOSOR14501-1G
APOSOR14501-250MG
3,4-Dihydro-2H-chromen-2-ylmethanol 95%
3,4-Dihydro-2H-chromen-2-ylmethanol
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658990 CAS-Nummer: 83278-86-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...