5-Bromo-1H-indazole-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR14502-1G OR14502-250MG OR14502-5G
APOSOR14502-1GEA 66.7 EUR
APOSOR14502-1G APOSOR14502-250MG APOSOR14502-5G
5-Bromo-1H-indazole-3-carboxylic acid
5-Bromo-1H-indazole-3-carboxylic acid
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241,0400 g/mol
Schmelzpunkt: 290…292 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663979
CAS-Nummer: 1077-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR