2-Amino-5-hydroxymethylpyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-pyridinemethanol, 6-Amino-3-pyridinemethanol, 2-Amino-5-(hydroxymethyl)pyridine, 2-Aminopyridine-5-methanol, (6-Aminopyridin-3-yl)methanol, (6-Amino-3-pyridyl)methanol

OR14503-1G OR14503-250MG
APOSOR14503-1GEA 65.4 EUR
APOSOR14503-1G APOSOR14503-250MG
2-Amino-5-hydroxymethylpyridine 97%
2-Amino-5-hydroxymethylpyridine
Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
Schmelzpunkt: 113…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659089
CAS-Nummer: 113293-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR