Methyl (2-chloromethyl)oxazole-4-carboxylate 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 2-(chloromethyl)oxazole-4-carboxylate

OR1452-1G OR1452-250MG
APOSOR1452-1GEA 62.5 EUR
APOSOR1452-1G APOSOR1452-250MG
Methyl (2-chloromethyl)oxazole-4-carboxylate 97%
Methyl (2-chloromethyl)oxazole-4-carboxylate
Formel: C₆H₆ClNO₃
Molecular Weight: 175,57 g/mol
Schmelzpunkt: 72…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660133
CAS-Nummer: 208465-72-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR