Boc-2-methyl-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: Boc-Phe(2-Me)-OH

OR14564-5G
APOSOR14564-5GEA 63.8 EUR
APOSOR14564-5G
Boc-2-methyl-L-phenylalanine 97%
Boc-2-methyl-L-phenylalanine
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279.34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671390
CAS-Nummer: 114873-05-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR