Fmoc-3-methyl-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(m-tolyl)propanoic acid, Fmoc-Phe(3-Me)-OH

OR14570-25G OR14570-5G
APOSOR14570-25GEA 543 EUR
APOSOR14570-25G APOSOR14570-5G
Fmoc-3-methyl-L-phenylalanine 97%
Fmoc-3-methyl-L-phenylalanine
Formel: C₂₅H₂₃NO₄
Molecular Weight: 401.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863050
CAS-Nummer: 211637-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR