Boc-3-nitro-L-phenylalanine

Lieferant: Apollo Scientific

Synonyme: Boc-L-3-Nitrophenylalanine, Boc-Phe(3-NO₂)-OH, 3-Nitro-L-phenylalanine, N-BOC protected

OR14580-1G OR14580-5G
APOSOR14580-1GEA 71.2 EUR
APOSOR14580-1G APOSOR14580-5G
Boc-3-nitro-L-phenylalanine
Boc-3-nitro-L-phenylalanine
Formel: C₁₄H₁₈N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD01317033
CAS-Nummer: 131980-29-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR