(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 98%

Lieferant: Apollo Scientific

Synonyme: Boc-Phe(4-NO2)-OH, Boc-Phe(4-NO₂)-OH, N-Boc-4-nitro-L-phenylalanine, Boc-p-nitro-Phe-OH

OR14584-100G
APOSOR14584-100GEA 74.1 EUR
APOSOR14584-100G
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid
Formel: C₁₄H₁₈N₂O₆
Molecular Weight: 310,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038128
CAS-Nummer: 33305-77-0
EINECS: 251-450-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR